


2-Butyne-1,4-diol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-65-6
:
Formula:C₄H₆O₂
MDL Number:MFCD00002915
:86.09 g/mol
Boiling Pt:238 °C (1013 hPa)
Melting Pt:55…58 °C
Density:1.1001 g/cm³ (20 °C)
Flash Pt:142 °C
Storage Temperature:Ambient
UN:2716
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:203-788-6
:GHS06
:GHS05
:GHS08