2-Butyne-1,4-diol 99%
:110-65-6
:
:C₄H₆O₂
:MFCD00002915
:86.09 g/mol
:238 °C (760 mmHg)
:54…58 °C
:1.2 g/cm³
:152 °C
:Ambient
:203-788-6
:2716
:6.1,III
:GHS05
:GHS06
:GHS08
- Appearance:Yellow to beige Crystals or flakes
- Infrared spectrum:Conforms
- Melting point:54 to 58 °C
- GC:≥98.5 %