Order Entry
Export
Orders LinkContactUsLinkComponent
 

3-Hydroxycinnamic acid

Glass bottle for solids, 5 g
Catalog # ROTH9904.1
Irritant
Supplier:   Roth Carl
3-Hydroxycinnamic acid
Catalog # ROTH9904.1
Supplier:  Roth Carl
CAS Number:  588-30-7
3-Hydroxycinnamic acid

Glass bottle for solids, 5 g

CAS Number:  588-30-7
Price
75,30 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  m-Hydroxycinnamic acid, 3-(3-Hydroxyphenyl)acrylic acid, m-Coumaric acid
  • CAS Number:588-30-7
  • Formula:C₉H₈O₃
  • MDL Number:MFCD00004390
  • Molecular Weight:164.158 g/mol
  • EINECS:209-615-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications