3,4-Dimethoxycinnamic acid, Sigma-Aldrich®
Catalog # SIALD133809-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:208.21 g/mol
Boiling Pt:343 °C (1013 hPa)
Melting Pt:198…202 °C
:M-Clarity™ quality level = MQ100
EINECS:219-025-5
CAS Number:2316-26-9
Formula:C₁₁H₁₂O₄
MDL Number:MFCD00004387