3-Chlorocinnamic acid
Glass bottle for solids, 25 g
Catalog # MOLEM22709700
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 3-(3-Chlorophenyl)acrylic acid, (2E)-3-(3-Chlorophenyl)acrylic acid
- CAS Number:1866-38-2
- Formula:C₉H₇ClO₂
- MDL Number:MFCD00004384
- Molecular Weight:182.61 g/mol
- EINECS:217-478-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



