2,3-Dimethyl-1-butene, Sigma-Aldrich®
SIAL190403-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:84.16 g/mol
:56 °C (1013 hPa)
:–157 °C
:0.6778 g/cm³ (20 °C)
:Ambient
:209-262-2
:3295
:3,II
:M-Clarity™ quality level = MQ200
:563-78-0
:C₆H₁₂
:MFCD00008923