Order Entry
Export
Orders LinkContactUsLinkComponent
Allyl alcohol

Allyl alcohol

0,5 g
Catalog # EHERC10130000
Flammable
Toxic
Environment
Supplier:   EHRENSTORFER
Allyl alcohol
Catalog # EHERC10130000
Supplier:  EHRENSTORFER
CAS Number:  107-18-6
Allyl alcohol

0,5 g

CAS Number:  107-18-6
Price
36,30 €
Log in for availability
1 * 0,5 g
Product Details & Documents
Synonyms:  3-Hydroxypropene, 2-Propen-1-ol
  • CAS Number:107-18-6
  • Formula:C₃H₆O
  • MDL Number:MFCD00002920
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00283
  • Boiling Pt:97 °C (1013 hPa)
  • Melting Pt:–129 °C
  • Density:0.854 g/cm³ (20 °C)
  • Flash Pt:21 °C
  • Storage Temperature:Refrigerator
  • EINECS:203-470-7
  • UN:1098
  • ADR:6.1,I
Specifications

Specifications

  • Pk:
    0,5 g

Specifications