
10-Undecen-1-ol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-43-6
:
Formula:C₁₁H₂₂O
MDL Number:MFCD00004750
Boiling Pt:132…133 °C (15 mmHg)
Melting Pt:–3 °C
Density:0.848
Flash Pt:93 °C (199 °F)
:170.3 g/mol
EINECS:203-971-0
:GHS07
Storage Temperature:Ambient