ContactUsLinkComponent
Fumaric acid
  EHERC13955500
 :  
Fumaric acid
  EHERC13955500
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029
  • Environmentally Preferable:
 : trans-1,2-Ethylentricarboxylic acid
 :
 :116.07 g/mol
 :13,04308
 :356 °C (1013 hPa)
 :287 °C
 :1.625 g/cm³ (20 °C)
 :273 °C
 :203-743-0
 :GHS07
 :110-17-8
 :C₄H₄O₄
 :MFCD00002700