Sodium oleate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:143-19-1
:
Formula:C₁₈H₃₃NaO₂
MDL Number:MFCD00004438
Melting Pt:232…235 °C
Density:1.02 g/cm³ (20 °C)
:304.45 g/mol
Storage Temperature:Freezer
EINECS:205-591-0