Oleic acid
Catalog # EHERC15727000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
Merck Index:13,06898
Boiling Pt:286 °C (1013 hPa)
Melting Pt:16.7 °C
Density:0.8946 g/cm³ (20 °C)
Flash Pt:180 °C
:282.47 g/mol
Storage Temperature:Freezer
EINECS:204-007-1
CAS Number:112-80-1
Formula:C₁₈H₃₄O₂
MDL Number:MFCD00064242