Order Entry
Export
ContactUsLinkComponent
 
Sodium oleate 65-90% oleic C18
Catalog # ACRO215080010
CAS Number:  
undefined
Sodium oleate 65-90% oleic C18
Catalog # ACRO215080010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • GC:
    ≥65 %

Specification Test Results

About this item

 :
Merck Index:13,06898
Boiling Pt:286 °C (1013 hPa)
Melting Pt:232…235 °C
Density:0,8946 g/cm³ (20 °C)
Flash Pt:180 °C
 :304.45 g/mol
Storage Temperature:Refrigerator
EINECS:204-007-1
CAS Number:143-19-1
Formula:C₁₈H₃₃NaO₂
MDL Number:MFCD00004438