Methyl oleate
Glass bottle for solids, 1 g
Catalog # ROTH3186.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester
- CAS Number:112-62-9
- Formula:C₁₉H₃₆O₂
- MDL Number:MFCD00009578
- Molecular Weight:296.49 g/mol
- Merck Index:12,06965
- Boiling Pt:∼218.5 °C (1013 hPa)
- Melting Pt:–20 °C
- Density:0.88 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:203-992-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



