Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bisacrylamide

Glass bottle for solids, 100 g
Catalog # MOLEM43851178
Irritant
Supplier:   Molekula
Bisacrylamide
Catalog # MOLEM43851178
Supplier:  Molekula
CAS Number:  110-26-9
Bisacrylamide
Bisacrylamide

Glass bottle for solids, 100 g

CAS Number:  110-26-9
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
  • CAS Number:110-26-9
  • Formula:(H₂C=CHCONH)₂CH₂
  • MDL Number:MFCD00008625
  • Molecular Weight:154.17 g/mol
  • Boiling Pt:125 °C (3.33 hPa)
  • Melting Pt:185 °C
  • Density:1.235 g/cm³ (20 °C)
  • Flash Pt:138 °C
  • Storage Temperature:Refrigerator
  • EINECS:203-750-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications