Order Entry
Export
Orders LinkContactUsLinkComponent
Acrylamide, 4x crystallised

Acrylamide, 4x crystallised

Toxic
Health
Acrylamide, 4x crystallised
Certificates
Acrylamide, 4x crystallised
Acrylamide, 4x crystallised

2 Options Available Below

Product Details & Documents
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Boiling Pt:232 °C (1013 hPa)
  • Melting Pt:84.5 °C
  • Density:1.122 g/cm³ (20 °C)
  • Flash Pt:138 °C
  • Storage Temperature:Refrigerator
  • EINECS:201-173-7
  • UN:2074
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g