Order Entry
Export
Orders LinkContactUsLinkComponent
Tetrachloroethylene

Tetrachloroethylene

Health
Environment
Supplier:   Roth Carl
Tetrachloroethylene
Supplier:  Roth Carl
Certificates
Tetrachloroethylene
Tetrachloroethylene

3 Options Available Below

Product Details & Documents
Synonyms:  Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
  • CAS Number:127-18-4
  • Formula:C₂Cl₄
  • MDL Number:MFCD00000834
  • Molecular Weight:165.83 g/mol
  • Merck Index:12,09332
  • Boiling Pt:121.1 °C (1013 hPa)
  • Melting Pt:–22 °C
  • Density:1.63 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-825-9
  • UN:1897
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 35,90 €
    35,90 €
    1 * 1 L
       35,90 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 65,30 €
    65,30 €
    1 * 2,5 L
       65,30 €

  • 10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 213,00 €
    213,00 €
    1 * 10 L
       213,00 €