Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
11-Bromo-1-undecene
11-Bromo-1-undecene, Sigma-Aldrich®
11-Bromo-1-undecene, Sigma-Aldrich®
Irritant
11-Bromo-1-undecene, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Irritant
11-Bromo-1-undecene, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
7766-50-9
:
Formula:
C₁₁H₂₁Br
MDL Number:
MFCD00040825
:
233.19 g/mol
:
M-Clarity™ quality level = MQ100
:
GHS07
Storage Temperature:
Ambient