11-Bromo-1-undecene ≥96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7766-50-9
:
Formula:C₁₁H₂₁Br
MDL Number:MFCD00040825
:233.19 g/mol
Boiling Pt:149…150 °C (35 mmHg)
Density:1.063
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient