HEPES sodium salt 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75277-39-3
:
Formula:C₈H₁₇N₂NaO₄S
MDL Number:MFCD00036463
:260.29 g/mol
Melting Pt:248 °C
Storage Temperature:Ambient
EINECS:278-169-7
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤3 % (K.F.)
- UV:(0.1 M in water)
- pH:9.5 to 10.5 (1 % in water)
- Titration with HClO4:≥98.5 % (on anhydrous substance)