
(2,4-Difluorophenyl)acetic acid, Sigma-Aldrich®
Catalog # SIAL264474-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:172.13 g/mol
Boiling Pt:259 °C (1013 hPa)
Melting Pt:117…119 °C
:M-Clarity™ quality level = MQ100
EINECS:279-709-4
:GHS07
CAS Number:81228-09-3
Formula:(C₆H₃F₂)CH₂COOH
MDL Number:MFCD00009999