
(2-Hydroxyphenyl)acetic acid 99%
:614-75-5
:
:(C₆H₄OH)CH₂COOH
:MFCD00004323
:152.15 g/mol
:328 °C (1013 hPa)
:143…148 °C
:Ambient
:210-393-2
:GHS07
- Appearance:Light pink to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:143 to 148 °C
- HPLC:≥98.5 %
- Titration with NaOH:98.5 to 101.5 %