
3-(3,4-Dimethoxyphenyl)propionic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2107-70-2
:
Formula:C₁₁H₁₄O₄
MDL Number:MFCD00002774
:210.23 g/mol
Boiling Pt:344 °C (1013 hPa)
Melting Pt:96…99 °C
Storage Temperature:Ambient
EINECS:218-288-3
:GHS07