Order Entry
Export
ContactUsLinkComponent
(3-Methoxyphenyl)acetic acid
Corrosive
(3-Methoxyphenyl)acetic acid
Catalog # MOLE21703278-25G
Supplier:  Molekula
CAS Number:  
Corrosive
(3-Methoxyphenyl)acetic acid
Catalog # MOLE21703278-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :166.18 g/mol
Boiling Pt:305.95 °C (1013 hPa)
Melting Pt:71…73 °C
EINECS:217-282-8
 :GHS05
Storage Temperature:Ambient
CAS Number:1798-09-0
Formula:(C₆H₄(OCH₃))CH₂COOH
MDL Number:MFCD00004334
Boiling Pt:305,95 °C (1013 hPa)