Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Indol-3-yl)propionic acid
3-(Indol-3-yl)propionic acid
3-(Indol-3-yl)propionic acid
3-(Indol-3-yl)propionic acid
Supplier:
Merck
Certificates
Expand image
3-(Indol-3-yl)propionic acid
Supplier:
Merck
About this item
CAS Number:
830-96-6
:
Formula:
C₁₁H₁₁NO₂
MDL Number:
MFCD00005660
:
189.21 g/mol
Melting Pt:
129…132 °C
Density:
1.356 g/cm³ (20 °C)
EINECS:
212-600-1