Order Entry
Export
ContactUsLinkComponent
3-(Indol-3-yl)propionic acid, Sigma-Aldrich®
3-(Indol-3-yl)propionic acid, Sigma-Aldrich®
Catalog # SIAL57400-5G-F
Supplier:  Merck
CAS Number:  
3-(Indol-3-yl)propionic acid, Sigma-Aldrich®
Catalog # SIAL57400-5G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :189.21 g/mol
Melting Pt:129…132 °C
Density:1.356 g/cm³ (20 °C)
EINECS:212-600-1
CAS Number:830-96-6
Formula:C₁₁H₁₁NO₂
MDL Number:MFCD00005660