3-(Indol-3-yl)propionic acid
Catalog # MOLE10186726-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:189.21 g/mol
Melting Pt:129…132 °C
Density:1.356 g/cm³ (20 °C)
EINECS:212-600-1
CAS Number:830-96-6
Formula:C₁₁H₁₁NO₂
MDL Number:MFCD00005660