2-Methylbutane
Glass bottle, 1 L
Catalog # ROTH3927.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: iso-Pentane
- CAS Number:78-78-4
- Formula:CH₃CH₂CH(CH₃)₂
- MDL Number:MFCD00009338
- Molecular Weight:72.15 g/mol
- Boiling Pt:28 °C (1013 hPa)
- Melting Pt:–160 °C
- Density:0.62 g/cm³ (20 °C)
- Flash Pt:–51 °C
- Storage Temperature:Ambient
- EINECS:201-142-8
- UN:1265
- ADR:3,I
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



