2,4-Dimethylhexane, Sigma-Aldrich®
SIAL110574-10G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
:
:114.23 g/mol
:109 °C (1013 hPa)
:–124 °C
:0.9372 g/cm³ (20 °C)
:Ambient
:209-649-6
:1262
:3,II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS08
:GHS07
:GHS09
:589-43-5
:C₈H₁₈
:MFCD00039929