



2,2-Dimethylpentane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:590-35-2
:
Formula:C₇H₁₆
MDL Number:MFCD00009404
:100.2 g/mol
Boiling Pt:79 °C (1013 hPa)
Melting Pt:–123.7 °C
Density:0.674 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-680-5
ADR:3,II
:M-Clarity™ quality level = MQ100
UN:1206
:GHS02
:GHS08
:GHS07
:GHS09