Order Entry
Export
Orders LinkContactUsLinkComponent
2,5-Dimethyl-2,5-hexanediol

2,5-Dimethyl-2,5-hexanediol

Irritant
Supplier:   CHEMPUR
2,5-Dimethyl-2,5-hexanediol
Supplier:  CHEMPUR
Certificates
2,5-Dimethyl-2,5-hexanediol

2 Options Available Below

Product Details & Documents
Synonyms:  1,1,4,4-Tetramethyl-1,4-butanediol
  • CAS Number:110-03-2
  • Formula:C₈H₁₈O₂
  • MDL Number:MFCD00004473
  • Molecular Weight:146.23 g/mol
  • Boiling Pt:214 °C (1013 hPa)
  • Melting Pt:85…88 °C
  • Density:0,898 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-731-5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

This product has been blocked by your organization. Please contact your purchasing department for more information