
2,2-Dimethyl-1,3-propanediol 99%
:126-30-7
:
:C₅H₁₂O₂
:MFCD00004685
:104.15 g/mol
:12,06547
:208 °C (760 mmHg)
:122…128 °C
:1,07 g/cm³ (20 °C)
:107 °C
:204-781-0
:GHS05
:Ambient
- Appearance:White Crystalline chunks
- Infrared spectrum:Conforms
- Melting point:123 to 127 °C
- Water:≤0.3 %
- GC:≥98.5 %
- Acid value:≤0.1 mg KOH/g