Order Entry
Export
ContactUsLinkComponent
tert-Butanol, Sigma-Aldrich®
tert-Butanol, Sigma-Aldrich®
Catalog # SIAL471712-100ML
Supplier:  Merck
CAS Number:  
undefined
tert-Butanol, Sigma-Aldrich®
Catalog # SIAL471712-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :74.12 g/mol
Merck Index:12,01577
Boiling Pt:82.2 °C (1013 hPa)
Melting Pt:25.3 °C
Density:0.7887 g/cm³ (20 °C)
Flash Pt:11 °C
EINECS:200-889-7
UN:1120
ADR:3,II
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:75-65-0
Formula:(CH₃)₃COH
MDL Number:MFCD00004464