

tert-Butanol
Catalog # ROTH6994.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:74.12 g/mol
Merck Index:12,01577
Boiling Pt:82.2 °C (1013 hPa)
Melting Pt:25.3 °C
Density:0.7887 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-889-7
UN:1120
ADR:3,II
:GHS02
:GHS07
CAS Number:75-65-0
Formula:(CH₃)₃COH
MDL Number:MFCD00004464