Isobutanol ≥99%
Supplier: Thermo Fisher Scientific
About this item
Purity: 99% CAS No.: 78-83-1 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: 28°(82°F) Refractive Index: 1.3960 Odour: Sweet pH: 7 Melting Point: -108° Boiling Point: 107-108° Beilstein Reference: 1730878 Merck Reference: 14,5131 UN No.: 1212 MDL No.: MFCD00004740
Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol
- CAS Number:78-83-1
- Formula:(CH₃)₂CHCH₂OH
- MDL Number:MFCD00004740
- Molecular Weight:74.12 g/mol
- Merck Index:14,5131
- Boiling Pt:107…108 °C
- Melting Pt:–108 °C
- Density:0.802
- Flash Pt:28 °C (82 °F)
- Storage Temperature:Ambient
- EINECS:201-148-0
- UN:1212
- ADR:3,III
2 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 78-83-1 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: 28°(82°F) Refractive Index: 1.3960 Odour: Sweet pH: 7 Melting Point: -108° Boiling Point: 107-108° Beilstein Reference: 1730878 Merck Reference: 14,5131 UN No.: 1212 MDL No.: MFCD00004740
Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol
- CAS Number:78-83-1
- Formula:(CH₃)₂CHCH₂OH
- MDL Number:MFCD00004740
- Molecular Weight:74.12 g/mol
- Merck Index:14,5131
- Boiling Pt:107…108 °C
- Melting Pt:–108 °C
- Density:0.802
- Flash Pt:28 °C (82 °F)
- Storage Temperature:Ambient
- EINECS:201-148-0
- UN:1212
- ADR:3,III
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- B23091.AP
- B23091.0F
- Environmentally Preferable:
- Safer Human and Environmental Health
- Environmental certification:
- Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.B23091.APEnvironmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by default and after discussionSafer Human and Environmental Health - Specifications:
More
Specifications
Cat. No.B23091.0FEnvironmentally PreferableEnvironmental certification



