
3,3-Dimethylbutyric acid, Sigma-Aldrich®
Catalog # SIALB88403-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:116.16 g/mol
Merck Index:13,01537
Boiling Pt:185 °C (1013 hPa)
Melting Pt:7 °C
Density:0.9303 g/cm³ (0 °C)
EINECS:213-982-2
UN:3265
ADR:8,III
:M-Clarity™ quality level = MQ100
:GHS05
CAS Number:1070-83-3
Formula:C₆H₁₂O₂
MDL Number:MFCD00002715