Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Propylvaleric acid

Glass bottle for solids, 500 mg
Catalog # PROOMM0751.00
2-Propylvaleric acid
Catalog # PROOMM0751.00
CAS Number:  99-66-1
2-Propylvaleric acid

Glass bottle for solids, 500 mg

CAS Number:  99-66-1
Price
1 320,00 €
Log in for availability
1 * 500 mg
Product Details & Documents
Synonyms:  2-Propylpentanoic acid, Valproic Acid, 2,2-Di-n-propylacetic acid
  • CAS Number:99-66-1
  • Formula:C₈H₁₆O₂
  • MDL Number:MFCD00002672
  • Molecular Weight:144.21 g/mol
  • Merck Index:12,10049
  • Boiling Pt:220 °C (1013 hPa)
  • Density:0.905 g/cm³ (20 °C)
  • Flash Pt:111 °C
  • Storage Temperature:Ambient
  • EINECS:202-777-3
  • UN:3265
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg

Specifications