
2,2-Dimethylcaproic acid ≥94%
Catalog # L14664.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:813-72-9
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00965097
:144.21 g/mol
Boiling Pt:216…220 °C
Density:0.913
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05