1-Chloro-2-methyl-2-propanol, Sigma-Aldrich®
Catalog # SIAL167592-50ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
:108.57 g/mol
Boiling Pt:124 °C (1013 hPa)
Melting Pt:–20 °C
Density:1.082 g/cm³ (0 °C)
Storage Temperature:Ambient
EINECS:209-196-4
UN:1987
ADR:3,III
:M-Clarity™ quality level = MQ100
CAS Number:558-42-9
Formula:C₄H₉ClO
MDL Number:MFCD00021807