n-Undecane, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Undecane
- CAS Number:1120-21-4
- Formula:H₃C(CH₂)₉CH₃
- MDL Number:MFCD00008959
- Molecular Weight:156.31 g/mol
- Boiling Pt:196 °C (1013 hPa)
- Melting Pt:–26.7 °C
- Density:0.7416 g/cm³ (20 °C)
- Flash Pt:60 °C
- Storage Temperature:Ambient
- EINECS:214-300-6
- ADR:3,III
- :M-Clarity™ quality level = MQ300
- UN:2330
3 Options Available Below
Product Details & Documents
Synonyms: Undecane
- CAS Number:1120-21-4
- Formula:H₃C(CH₂)₉CH₃
- MDL Number:MFCD00008959
- Molecular Weight:156.31 g/mol
- Boiling Pt:196 °C (1013 hPa)
- Melting Pt:–26.7 °C
- Density:0.7416 g/cm³ (20 °C)
- Flash Pt:60 °C
- Storage Temperature:Ambient
- EINECS:214-300-6
- ADR:3,III
- :M-Clarity™ quality level = MQ300
- UN:2330



