n-Undecane, Sigma-Aldrich®
Glass bottle, 25 mL
SIALU407-25ML
: Merck
: Undecane
- :1120-21-4
- :H₃C(CH₂)₉CH₃
- :MFCD00008959
- :156.31 g/mol
- :196 °C (1013 hPa)
- :–26.7 °C
- :0.7416 g/cm³ (20 °C)
- :60 °C
- :Ambient
- :214-300-6
- :3,III
- :M-Clarity™ quality level = MQ100
- :2330
- Pack type:Glass bottle
- Pk:25 mL