


EDTA tetrasodium salt dihydrate 99-101%, Ultrapure
:10378-23-1
:
:C₁₀H₁₂N₂Na₄O₈·2H₂O
:MFCD00150026
:416.2 g/mol
:GHS05
:GHS07
:GHS08
:Ambient
- Form:White crystalline powder
- Assay (unspecified):99 to 101% (as is)
- Identification (FTIR):Conforms
- pH (1%, H2O):10.7 to 11.7
- pH (5%, H2O):10.7 to 12.0