
n-Tetradecane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:629-59-4
:
Formula:H₃C(CH₂)₁₂CH₃
MDL Number:MFCD00008986
:198.39 g/mol
Boiling Pt:253.7 °C (1013 hPa)
Melting Pt:6 °C
Density:0.7628 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:102 °C (closed cup)
EINECS:211-096-0
:GHS08
Storage Temperature:Ambient