NucleoSpin® RNA kit
Supplier: MACHEREY-NAGEL
Certificates
About this item
NucleoSpin® RNA mini spin kit for the isolation of RNA of the highest integrity.
- rDNase included for on-column DNA digestion– easy and efficient removal of contaminating DNA
- NucleoSpin® filters (shredders) – efficient sample homogenization and reduction of viscosity
- Up to 70 μg ready to use RNA
- Parallel purification of genomic DNA by using the NucleoSpin® RNA/DNA buffer set
Delivery: Kit includes lysis buffer RA1, wash buffer RAW2, wash buffer RA3 (concentrate), membrane desalting buffer, reaction buffer for rDNase, rDNase, RNase-free (lyophilised), RNase-free H2O, NucleoSpin® filters (violet rings), NucleoSpin® RNA columns (light blue rings – plus collection tubes), collection tubes (2 ml), collection tubes (1,5 ml) and user manual.