

Methanesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:124-63-0
:
Formula:CH₃ClO₂S
MDL Number:MFCD00007454
:114.55 g/mol
Merck Index:13,05982
Boiling Pt:161 °C (1013 hPa)
Melting Pt:–32 °C
Density:1.478 g/cm³ (20 °C)
EINECS:204-706-1
UN:3246
ADR:6.1,I
:M-Clarity™ quality level = MQ200
Flash Pt:92 °C
:GHS06
:GHS05
Storage Temperature:Ambient