

Methanesulphonyl chloride 99.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:124-63-0
:
Formula:CH₃ClO₂S
MDL Number:MFCD00007454
:114.55 g/mol
Merck Index:13,05982
Boiling Pt:161 °C (1013 hPa)
Melting Pt:–33 °C
Density:1.48 g/cm³
Flash Pt:110 °C
Storage Temperature:Ambient
EINECS:204-706-1
UN:3246
ADR:6.1,I
:GHS06
:GHS05
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4510 to 1.4530 (20°C, 589 nm)
- Water:≤0.025 %
- GC:≥99.4 %
- Color scale:≤100 APHA