1,6-Hexanedithiol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1191-43-1
:
Formula:C₆H₁₄S₂
MDL Number:MFCD00004910
Boiling Pt:242 °C (1013 hPa)
Melting Pt:–21 °C
Density:1.0102 g/cm³ (0 °C)
ADR:6.1,III
:M-Clarity™ quality level = MQ300
:150.31 g/mol
Storage Temperature:Ambient
EINECS:214-735-1
UN:2810