1,5-Pentanedithiol, Sigma-Aldrich®
Catalog # SIAL242551-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:136.28 g/mol
Boiling Pt:229 °C (1013 hPa)
Melting Pt:–68.5 °C
Density:1.016 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:213-194-9
CAS Number:928-98-3
Formula:C₅H₁₂S₂
MDL Number:MFCD00004908