
1,3-Propanedithiol, Sigma-Aldrich®
Catalog # SIALP50609-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:108.23 g/mol
Merck Index:12,07984
Boiling Pt:169 °C (1013 hPa)
Melting Pt:–79 °C
Density:1.08 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-706-9
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:GHS07
CAS Number:109-80-8
Formula:C₃H₈S₂
MDL Number:MFCD00004904