1,2-Ethanedithiol, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: 1.2-Ethanedithiol, 1,2-Dimercaptoethane
- CAS Number:540-63-6
- Formula:C₂H₆S₂
- MDL Number:MFCD00004892
- Molecular Weight:94.2 g/mol
- Merck Index:13,03760
- Boiling Pt:146…149 °C (1013 hPa)
- Melting Pt:–41 °C
- Density:1.374 g/cm³ (20 °C)
- Flash Pt:44 °C
- Storage Temperature:Ambient
- UN:3071
- ADR:6.1,II
- :M-Clarity™ quality level = MQ200
- EINECS:208-752-3
2 Options Available Below
Product Details & Documents
Synonyms: 1.2-Ethanedithiol, 1,2-Dimercaptoethane
- CAS Number:540-63-6
- Formula:C₂H₆S₂
- MDL Number:MFCD00004892
- Molecular Weight:94.2 g/mol
- Merck Index:13,03760
- Boiling Pt:146…149 °C (1013 hPa)
- Melting Pt:–41 °C
- Density:1.374 g/cm³ (20 °C)
- Flash Pt:44 °C
- Storage Temperature:Ambient
- UN:3071
- ADR:6.1,II
- :M-Clarity™ quality level = MQ200
- EINECS:208-752-3



