1,7-Heptanediol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:629-30-1
:
Formula:C₇H₁₆O₂
MDL Number:MFCD00002987
:132.2 g/mol
Boiling Pt:259 °C (1013 hPa)
Melting Pt:17…18 °C
Density:0.953 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:211-085-0